Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 31, 32 | 3625 | ||
XM_005264254.1 | XP_005264311.1 | 31, 32 | 3456 | |||
XM_006711986.3 | XP_006712049.1 | 32, 33 | 3717 | |||
XM_006711987.1 | XP_006712050.1 | 31, 32 | 3522 | |||
XM_011532757.2 | XP_011531059.1 | 31, 32 | 3831 | |||
XM_011532758.1 | XP_011531060.1 | 31, 32 | 3522 | |||
XM_011532759.2 | XP_011531061.1 | 17, 18 | 2107 | |||
XM_011532760.2 | XP_011531062.1 | 14, 15 | 1787 | |||
XM_017003790.1 | XP_016859279.1 | 32, 33 | 3651 | |||
XM_017003791.1 | XP_016859280.1 | 30, 31 | 3592 | |||
XM_017003792.1 | XP_016859281.1 | 31, 32 | 3522 | |||
XM_017003793.1 | XP_016859282.1 | 27, 28 | 3032 | |||
XM_017003794.1 | XP_016859283.1 | 25, 26 | 2735 | |||
XM_017003795.1 | XP_016859284.1 | 29, 30 | 3584 | |||
GenBank mRNA | AB033005.1 | - | 13, 14 | 1490 | ||
AL110218.1 | - | 28, 29 | 3181 | |||
BC137126.1 | - | 31, 32 | 3515 | 9021123 |