Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 16 | 2624 | ||
NM_001288608.1 | NP_001275537.1 | 16 | 2810 | |||
NM_019032.5 | NP_061905.2 | 15 | 2741 | |||
NM_025008.4 | NP_079284.2 | 15 | 2741 | |||
XM_011509644.2 | XP_011507946.1 | 15 | 2772 | |||
XM_011509645.2 | XP_011507947.1 | 14 | 2700 | |||
XM_011509648.2 | XP_011507950.1 | 15 | 2726 | |||
XM_011509649.2 | XP_011507951.1 | 15 | 3016 | |||
XM_011509650.2 | XP_011507952.1 | 15 | 3013 | |||
XM_011509651.2 | XP_011507953.1 | 9 | 1522 | |||
XM_011509652.1 | XP_011507954.1 | 9 | 1228 | |||
XM_017001506.1 | XP_016856995.1 | 16 | 3181 | |||
XM_017001507.1 | XP_016856996.1 | 7 | 963 |