Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 19, 20 | 3268 | ||
NM_001288608.1 | NP_001275537.1 | 19, 20 | 3454 | |||
NM_019032.5 | NP_061905.2 | 18, 19 | 3385 | |||
XM_011509644.2 | XP_011507946.1 | 18, 19 | 3416 | |||
XM_011509645.2 | XP_011507947.1 | 17, 18 | 3344 | |||
XM_011509648.2 | XP_011507950.1 | 18, 19 | 3370 | |||
XM_011509651.2 | XP_011507953.1 | 12, 13 | 2166 | |||
XM_011509652.1 | XP_011507954.1 | 12, 13 | 1872 | |||
XM_017001506.1 | XP_016856995.1 | 19, 20 | 3825 | |||
XM_017001507.1 | XP_016856996.1 | 10, 11 | 1607 | |||
GenBank mRNA | AF217974.1 | - | 7, 8 | 1186 | ||
AF370411.1 | - | 12, 13 | 1922 | |||
AK293037.1 | - | 8, 9 | 1317 | |||
AL122085.1 | - | 2, 3 | 208 | |||
BC027478.1 | - | 11, 12 | 1824 | |||
BC071852.1 | - | 10, 11 | 1681 | |||
BC094811.1 | - | 10, 11 | 1681 | |||
BC140800.1 | - | 18, 19 | 3306 | 9021668 | ||
BC144581.1 | - | 18, 19 | 3120 | 9053111 | ||
BX647294.1 | - | 5, 6 | 947 |