Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172643.1 | NP_001166114.1 | 10 | 2386 | ||
NM_001172644.1 | NP_001166115.1 | 9 | 2747 | |||
NM_001172645.1 | NP_001166116.1 | 9 | 2768 | |||
NM_001271934.1 | NP_001258863.1 | 11 | 2917 | |||
NM_001271935.1 | NP_001258864.1 | 8 | 2217 | |||
NM_001272050.1 | NP_001258979.1 | 10 | 2596 | |||
NM_004614.4 | NP_004605.4 | 10 | 2822 | |||
NR_073520.1 | 9 | 3750 | ||||
GenBank mRNA | AK026041.1 | - | 1 | 742 | ||
BC040667.1 | - | 1 | 1171 |