Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000751.2 | NP_000742.1 | 11 | 1410 | ||
NM_001256657.1 | NP_001243586.1 | 10 | 1365 | |||
NM_001311195.1 | NP_001298124.1 | 9 | 1099 | |||
NM_001311196.1 | NP_001298125.1 | 11 | 1378 | |||
XM_011510524.2 | XP_011508826.1 | 10 | 1272 | |||
GenBank mRNA | AK291526.1 | - | 11 | 1382 | ||
AK300109.1 | - | 10 | 1365 | |||
AK304890.1 | - | 9 | 1071 | |||
AK315297.1 | - | 11 | 1382 | |||
BC093923.1 | - | 11 | 1386 | 7939768 | ||
BC093925.1 | - | 11 | 1386 | 7939770 | ||
CD013895.1 | - | 11 | 1323 | |||
X55019.1 | - | 11 | 1358 |