Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 8 | 1427 | ||
NM_007325.4 | NP_015564.4 | 8 | 1427 | |||
GenBank mRNA | AK297136.1 | - | 8 | 1113 | ||
BC117464.1 | - | 8 | 1299 | 40126015 | ||
DA180344.1 | - | 1 | 48 | |||
U10301.1 | - | 8 | 1187 | |||
U10302.1 | - | 8 | 1187 |