Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015166.3 | NP_055981.1 | 5 | 1006 | ||
NM_139202.2 | NP_631941.1 | 5 | 645 | |||
XM_011530678.1 | XP_011528980.1 | 5 | 767 | |||
XM_017028671.1 | XP_016884160.1 | 4 | 434 | |||
GenBank mRNA | AF319633.1 | - | 5 | 568 | ||
AK124264.1 | - | 5 | 1007 | |||
AK295106.1 | - | 4 | 267 | |||
AK299841.1 | - | 4 | 414 | |||
BC028425.2 | - | 5 | 605 | 4817175 | ||
BC070042.1 | - | 5 | 603 | 5285102 | ||
CR456460.1 | - | 4 | 424 | |||
D25217.3 | - | 5 | 682 |