Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000147.4 | NP_000138.2 | 8 | 1454 | ||
XM_005245821.2 | XP_005245878.1 | 8 | 1591 | |||
XM_011541167.2 | XP_011539469.1 | 8 | 1039 | |||
XM_017000905.1 | XP_016856394.1 | 8 | 1136 | |||
GenBank mRNA | AB074175.1 | - | 8 | 1375 | ||
AK092914.1 | - | 9 | 1309 | |||
AK095443.1 | - | 9 | 1326 | |||
AK095455.1 | - | 9 | 1340 | |||
AK225468.1 | - | 8 | 1375 | |||
AK301302.1 | - | 10 | 1304 | |||
AK314649.1 | - | 8 | 1375 | |||
BC017338.2 | - | 8 | 1377 | 4871788 | ||
M29877.1 | - | 8 | 1374 |