Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277176.1 | NP_001264105.1 | 3 | 832 | ||
NM_182758.3 | NP_877435.3 | 20 | 3893 | |||
NR_102334.1 | 20 | 4091 | ||||
NR_102335.1 | 3 | 917 | ||||
NR_102336.1 | 3 | 894 | ||||
XM_011521433.2 | XP_011519735.1 | 21 | 4090 | |||
XM_011521435.2 | XP_011519737.1 | 20 | 3944 | |||
XM_017022061.1 | XP_016877550.1 | 20 | 3888 | |||
GenBank mRNA | AK123805.1 | - | 1 | 466 | ||
AK125130.1 | - | 6 | 1504 | |||
AK310211.1 | - | 3 | 917 | |||
AK311300.1 | - | 3 | 831 | |||
BX537884.1 | - | 20 | 3893 | |||
BX648571.1 | - | 20 | 4093 | |||
BX648751.1 | - | 14 | 3274 |