Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277176.1 | NP_001264105.1 | 3 | 286 | ||
NM_182758.3 | NP_877435.3 | 20 | 3347 | |||
NR_102334.1 | 20 | 3545 | ||||
NR_102335.1 | 3 | 371 | ||||
NR_102336.1 | 3 | 348 | ||||
XM_011521433.2 | XP_011519735.1 | 21 | 3544 | |||
XM_011521435.2 | XP_011519737.1 | 20 | 3398 | |||
XM_011521436.2 | XP_011519738.1 | 20 | 3373 | |||
XM_011521437.2 | XP_011519739.1 | 19 | 3271 | |||
XM_017022061.1 | XP_016877550.1 | 20 | 3342 | |||
GenBank mRNA | AK125130.1 | - | 6 | 958 | ||
AK310211.1 | - | 3 | 371 | |||
AK311300.1 | - | 3 | 285 | |||
BC101614.1 | - | 19 | 3326 | 8069120 | ||
BC101616.1 | - | 19 | 3326 | 8069122 | ||
BX537884.1 | - | 20 | 3347 | |||
BX648571.1 | - | 20 | 3547 | |||
BX648751.1 | - | 14 | 2728 | |||
DA736178.1 | - | 3 | 355 | |||
DA748816.1 | - | 3 | 278 |