Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001710.5 | NP_001701.2 | 13 | 1960 | ||
GenBank mRNA | AK130533.1 | - | 13 | 1889 | ||
AK223400.1 | - | 13 | 1808 | |||
BC004143.2 | - | 13 | 1817 | 2959705 | ||
BC007990.2 | - | 13 | 1817 | 2959706 | ||
K01566.1 | - | 12 | 1421 | |||
L15702.1 | - | 13 | 1722 | |||
S67310.1 | - | 13 | 1793 | |||
X72875.1 | - | 13 | 1682 |