Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001710.5 | NP_001701.2 | 17 | 2404 | ||
GenBank mRNA | AK130533.1 | - | 17 | 2333 | ||
AK223400.1 | - | 17 | 2252 | |||
BC004143.2 | - | 17 | 2261 | 2959705 | ||
BC007990.2 | - | 17 | 2261 | 2959706 | ||
L15702.1 | - | 17 | 2166 | |||
S67310.1 | - | 17 | 2237 | |||
X72875.1 | - | 17 | 2126 |