Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122957.2 | NP_001116429.1 | 6 | 893 | ||
NM_001271926.1 | NP_001258855.1 | 6 | 893 | |||
NM_005881.3 | NP_005872.2 | 6 | 893 | |||
XM_017022859.1 | XP_016878348.1 | 6 | 806 | |||
GenBank mRNA | AF026548.1 | - | 6 | 803 | ||
AK130145.1 | - | 6 | 773 | |||
AK294691.1 | - | 5 | 602 | |||
BC007363.2 | - | 6 | 776 | 3630050 | ||
BC009872.2 | - | 6 | 756 | 3944953 | ||
CR542093.1 | - | 5 | 530 |