Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000285.3 | NP_000276.2 | 12 | 1028 | ||
NM_001166056.1 | NP_001159528.1 | 10 | 905 | |||
NM_001166057.1 | NP_001159529.1 | 10 | 836 | |||
GenBank mRNA | AK225037.1 | - | 12 | 909 | ||
AK290756.1 | - | 12 | 909 | |||
AK290781.1 | - | 12 | 924 | |||
AK291561.1 | - | 12 | 926 | |||
AK293126.1 | - | 10 | 731 | |||
AK294619.1 | - | 10 | 803 | |||
BC004305.1 | - | 12 | 899 | 3626134 | ||
BC015027.1 | - | 12 | 929 | 3920275 | ||
BC028295.1 | - | 12 | 900 | 4470066 | ||
BT006692.1 | - | 12 | 895 | |||
CR541669.1 | - | 12 | 895 | |||
J04605.1 | - | 12 | 911 |