Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260492.1 | NP_001247421.1 | 14 | 2043 | ||
NM_001260493.1 | NP_001247422.1 | 14 | 2043 | |||
NM_001260494.1 | NP_001247423.1 | 12 | 1672 | |||
NM_001260495.1 | NP_001247424.1 | 7 | 1096 | |||
NM_001260496.1 | NP_001247425.1 | 6 | 831 | |||
NM_002906.3 | NP_002897.1 | 14 | 2043 | |||
GenBank mRNA | AK295075.1 | - | 11 | 1388 | ||
AK312903.1 | - | 14 | 1945 | |||
AK316061.1 | - | 12 | 1574 | |||
BC047109.1 | - | 14 | 1943 | 5284438 | ||
BX648536.1 | - | 4 | 759 | |||
DQ916738.1 | - | 14 | 2043 | |||
DQ916739.1 | - | 6 | 831 | |||
DQ916740.1 | - | 7 | 1096 | |||
DQ916741.1 | - | 14 | 2043 | |||
DQ916742.1 | - | 14 | 2043 | |||
L02320.1 | - | 13 | 1763 |