Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006765.3 | NP_006756.2 | 9, 10 | 1366 | ||
XM_011544652.2 | XP_011542954.1 | 8, 9 | 909 | |||
XM_017013861.1 | XP_016869350.1 | 8, 9 | 909 | |||
GenBank mRNA | AY562998.1 | - | 2, 3 | 125 | ||
BT020002.1 | - | 9, 10 | 1022 | |||
U42349.1 | - | 9, 10 | 1179 |