Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178234.2 | NP_839952.1 | 9, 10 | 1366 | ||
GenBank mRNA | BC010370.1 | - | 9, 10 | 1234 | 4334284 |