Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003384.2 | NP_003375.1 | 9 | 834 | ||
XM_006720247.3 | XP_006720310.1 | 9 | 872 | |||
XM_011537132.1 | XP_011535434.1 | 8 | 733 | |||
XM_017021622.1 | XP_016877111.1 | 8 | 733 | |||
XM_017021624.1 | XP_016877113.1 | 9 | 872 | |||
XM_017021625.1 | XP_016877114.1 | 9 | 913 | |||
GenBank mRNA | AB000449.1 | - | 9 | 803 | ||
AK290110.1 | - | 9 | 822 | |||
BC103761.1 | - | 9 | 812 | 5773877 | ||
BC112075.1 | - | 9 | 802 | 8327543 | ||
BC113510.1 | - | 9 | 802 | 8322562 |