Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 22 | 3596 | ||
NM_001040716.1 | NP_001035806.1 | 23 | 3784 | |||
NM_022172.2 | NP_071504.2 | 21 | 3551 | |||
XM_005274031.4 | XP_005274088.1 | 21 | 3643 | |||
XM_005274032.4 | XP_005274089.1 | 21 | 3525 | |||
XM_006718578.3 | XP_006718641.1 | 21 | 3596 | |||
XM_006718579.3 | XP_006718642.1 | 13 | 2385 | |||
XM_011545086.2 | XP_011543388.1 | 21 | 3912 | |||
XM_011545087.2 | XP_011543389.1 | 13 | 2510 | |||
XM_017017868.1 | XP_016873357.1 | 24 | 4083 | |||
XM_017017869.1 | XP_016873358.1 | 24 | 3954 | |||
XM_017017870.1 | XP_016873359.1 | 23 | 3895 | |||
XM_017017871.1 | XP_016873360.1 | 23 | 3766 | |||
XM_017017872.1 | XP_016873361.1 | 22 | 4148 | |||
GenBank mRNA | BC011617.2 | - | 22 | 3585 | 3946441 | |
K02282.1 | - | 1 | 101 | |||
M26122.1 | - | 2 | 256 | |||
S72370.1 | - | 21 | 3584 | |||
U04641.1 | - | 21 | 3540 | |||
U30891.1 | - | 22 | 3554 |