Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033503.2 | NP_001028675.1 | 6 | 577 | ||
NM_016103.3 | NP_057187.1 | 5 | 458 | |||
XM_017009521.1 | XP_016865010.1 | 5 | 465 | |||
GenBank mRNA | AF087850.1 | - | 5 | 495 | ||
AF092130.1 | - | 5 | 474 | |||
AK056821.1 | - | 2 | 2292 | |||
AK223452.1 | - | 5 | 443 | |||
AL512710.1 | - | 5 | 1493 | |||
BC002847.2 | - | 6 | 508 | 3636540 | ||
BC093034.1 | - | 5 | 458 | 30528518 | ||
CD048852.1 | - | 6 | 585 |