Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033503.2 | NP_001028675.1 | 7 | 630 | ||
NM_016103.3 | NP_057187.1 | 6 | 511 | |||
XM_017009521.1 | XP_016865010.1 | 6 | 518 | |||
GenBank mRNA | AF087850.1 | - | 6 | 548 | ||
AF092130.1 | - | 6 | 527 | |||
AK056821.1 | - | 3 | 2345 | |||
AK223452.1 | - | 6 | 496 | |||
AL512710.1 | - | 6 | 1546 | |||
BC002847.2 | - | 7 | 561 | 3636540 | ||
BC093034.1 | - | 6 | 511 | 30528518 | ||
CD048852.1 | - | 7 | 638 |