Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160708.1 | NP_001154180.1 | 9 | 735 | ||
NM_018082.5 | NP_060552.4 | 9 | 888 | |||
XM_017019621.1 | XP_016875110.1 | 9 | 853 | |||
GenBank mRNA | AK122713.1 | - | 9 | 715 | ||
AK291635.1 | - | 9 | 732 | |||
AY092084.1 | - | 9 | 719 | |||
BC046238.1 | - | 9 | 755 | 4905794 | ||
BX537447.1 | - | 9 | 836 |