Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160708.1 | NP_001154180.1 | 21 | 2426 | ||
NM_018082.5 | NP_060552.4 | 21 | 2579 | |||
XM_017019621.1 | XP_016875110.1 | 21 | 2544 | |||
GenBank mRNA | AK001161.1 | - | 1 | 50 | ||
AK001250.1 | - | 2 | 108 | |||
AK090636.1 | - | 6 | 651 | |||
AK092031.1 | - | 11 | 1502 | |||
AK122713.1 | - | 21 | 2406 | |||
AK291635.1 | - | 21 | 2423 | |||
AY092084.1 | - | 21 | 2410 | |||
BC046238.1 | - | 21 | 2446 | 4905794 | ||
BX537447.1 | - | 21 | 2527 |