Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109878.1 | NP_001103348.1 | 4, 5 | 601 | ||
NM_001109879.1 | NP_001103349.1 | 4, 5 | 597 | |||
NM_001303475.1 | NP_001290404.1 | 2, 3 | 355 | |||
NM_016954.2 | NP_058650.1 | 3, 4 | 494 | |||
XM_005262136.2 | XP_005262193.1 | 4, 5 | 629 | |||
XM_006724657.3 | XP_006724720.1 | 4, 5 | 630 | |||
XM_011530972.1 | XP_011529274.1 | 3, 4 | 1234 | |||
GenBank mRNA | AF251684.1 | - | 2, 3 | 346 | ||
AK097514.1 | - | 4, 5 | 597 | |||
AY035371.1 | - | 3, 4 | 473 | |||
BC014194.2 | - | 3, 4 | 494 | 3952809 |