Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000299.3 | NP_000290.2 | 3 | 799 | ||
NM_001005337.2 | NP_001005337.1 | 3 | 799 | |||
GenBank mRNA | AK315291.1 | - | 3 | 745 | ||
BC114571.1 | - | 3 | 682 | 40082609 | ||
X79293.1 | - | 3 | 739 | |||
Z34974.1 | - | 3 | 800 |