Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 10 | 1753 | ||
NM_001284290.1 | NP_001271219.1 | 8 | 1315 | |||
NM_001293104.1 | NP_001280033.1 | 9 | 1568 | |||
NM_001293105.1 | NP_001280034.1 | 8 | 1425 | |||
NR_120531.1 | 9 | 1668 | ||||
XM_005250297.3 | XP_005250354.1 | 9 | 1595 | |||
XM_011516113.1 | XP_011514415.1 | 9 | 1497 | |||
XM_011516114.1 | XP_011514416.1 | 9 | 1425 | |||
XM_017012091.1 | XP_016867580.1 | 8 | 1344 | |||
XM_017012092.1 | XP_016867581.1 | 8 | 1331 | |||
XM_017012093.1 | XP_016867582.1 | 8 | 1272 | |||
GenBank mRNA | AK096764.1 | - | 2 | 2011 | ||
AK223406.1 | - | 10 | 1652 | |||
AK300223.1 | - | 10 | 1542 | |||
AK303819.1 | - | 8 | 1212 | |||
AK312752.1 | - | 10 | 1669 | |||
BC014142.2 | - | 10 | 1623 | 4662011 | ||
CD014092.1 | - | 9 | 1450 | |||
CD014093.1 | - | 8 | 1308 | |||
CD014094.1 | - | 9 | 1550 | |||
M10618.1 | - | 1 | 65 | |||
M15182.1 | - | 10 | 1648 |