Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278682.1 | NP_001265611.1 | 3 | 763 | ||
NM_001278684.1 | NP_001265613.1 | 4 | 1120 | |||
NM_001278686.1 | NP_001265615.1 | 4 | 786 | |||
NM_003244.3 | NP_003235.1 | 3 | 853 | |||
NM_170695.3 | NP_733796.2 | 3 | 1497 | |||
NM_173207.2 | NP_775299.1 | 3 | 731 | |||
NM_173208.2 | NP_775300.1 | 4 | 1123 | |||
NM_173209.2 | NP_775301.1 | 3 | 939 | |||
NM_173210.2 | NP_775302.1 | 3 | 890 | |||
NM_173211.1 | NP_775303.1 | 3 | 602 | |||
NM_174886.2 | NP_777480.1 | 4 | 920 | |||
XM_011525735.2 | XP_011524037.1 | 3 | 1099 | |||
XM_017025958.1 | XP_016881447.1 | 4 | 1184 | |||
XM_017025959.1 | XP_016881448.1 | 3 | 969 | |||
GenBank mRNA | AF179900.1 | - | 3 | 541 | ||
AK026766.1 | - | 2 | 1459 | |||
AK291112.1 | - | 3 | 687 | |||
BC000814.1 | - | 3 | 619 | 3453337 | ||
BC031268.1 | - | 3 | 1225 | 5284849 | ||
CR450333.1 | - | 3 | 535 | |||
X89750.1 | - | 3 | 846 |