Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278682.1 | NP_001265611.1 | 3 | 868 | ||
NM_001278684.1 | NP_001265613.1 | 4 | 1225 | |||
NM_001278686.1 | NP_001265615.1 | 4 | 891 | |||
NM_003244.3 | NP_003235.1 | 3 | 958 | |||
NM_170695.3 | NP_733796.2 | 3 | 1602 | |||
NM_173207.2 | NP_775299.1 | 3 | 836 | |||
NM_173208.2 | NP_775300.1 | 4 | 1228 | |||
NM_173209.2 | NP_775301.1 | 3 | 1044 | |||
NM_173210.2 | NP_775302.1 | 3 | 995 | |||
NM_173211.1 | NP_775303.1 | 3 | 707 | |||
NM_174886.2 | NP_777480.1 | 4 | 1025 | |||
XM_011525735.2 | XP_011524037.1 | 3 | 1204 | |||
XM_017025958.1 | XP_016881447.1 | 4 | 1289 | |||
XM_017025959.1 | XP_016881448.1 | 3 | 1074 | |||
GenBank mRNA | AF179900.1 | - | 3 | 646 | ||
AK026766.1 | - | 2 | 1564 | |||
AK291112.1 | - | 3 | 792 | |||
BC000814.1 | - | 3 | 724 | 3453337 | ||
BC031268.1 | - | 3 | 1330 | 5284849 | ||
CR450333.1 | - | 3 | 640 | |||
X89750.1 | - | 3 | 951 |