Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 27 | 9100 | ||
NM_005933.3 | NP_005924.2 | 27 | 9091 | |||
XM_006718839.3 | XP_006718902.2 | 27 | 6575 | |||
XM_011542829.2 | XP_011541131.1 | 28 | 9191 | |||
XM_011542830.2 | XP_011541132.1 | 28 | 9188 | |||
XM_011542831.2 | XP_011541133.1 | 28 | 9182 | |||
XM_011542833.2 | XP_011541135.1 | 28 | 6674 | |||
GenBank mRNA | AB209508.1 | - | 25 | 6635 | ||
AK303179.1 | - | 1 | 565 | |||
L04284.1 | - | 27 | 9068 | |||
L04731.1 | - | 27 | 8891 |