Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 27 | 10638 | ||
NM_005933.3 | NP_005924.2 | 27 | 10629 | |||
XM_006718839.3 | XP_006718902.2 | 27 | 8113 | |||
XM_011542829.2 | XP_011541131.1 | 28 | 10729 | |||
XM_011542830.2 | XP_011541132.1 | 28 | 10726 | |||
XM_011542831.2 | XP_011541133.1 | 28 | 10720 | |||
XM_011542833.2 | XP_011541135.1 | 28 | 8212 | |||
GenBank mRNA | AB209508.1 | - | 25 | 8173 | ||
AK303179.1 | - | 1 | 2103 | |||
L04284.1 | - | 27 | 10606 | |||
L04731.1 | - | 27 | 10429 |