Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213606.3 | NP_998771.3 | 6 | 1032 | ||
XM_017016237.1 | XP_016871726.1 | 5 | 1122 | |||
XM_017016238.1 | XP_016871727.1 | 5 | 1061 | |||
XM_017016239.1 | XP_016871728.1 | 5 | 920 | |||
GenBank mRNA | AK124901.1 | - | 2 | 311 | ||
AK127303.1 | - | 6 | 1033 | |||
CR936657.1 | - | 5 | 891 |