Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009611.4 | NP_001009611.2 | 2 | 340 | ||
GenBank mRNA | AL049681.1 | - | 2 | 340 |