Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282489.2 | NP_001269418.1 | 3 | 2299 | ||
NM_173477.4 | NP_775748.2 | 3 | 2352 | |||
XM_011524296.2 | XP_011522598.1 | 3 | 2325 | |||
GenBank mRNA | AK091243.1 | - | 3 | 2345 |