Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006767.3 | NP_006758.2 | 11, 12 | 1369 | ||
GenBank mRNA | AK296347.1 | - | 10, 11 | 1250 | ||
AK310747.1 | - | 9, 10 | 972 | |||
AK313502.1 | - | 6, 7 | 771 | |||
BC026214.1 | - | 11, 12 | 1271 | 4399716 | ||
CT841521.1 | - | 11, 12 | 1285 | |||
D38496.1 | - | 11, 12 | 1264 |