Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079.3 | NP_001070.2 | 5 | 829 | ||
XM_017004867.1 | XP_016860356.1 | 7 | 2230 | |||
XM_017004868.1 | XP_016860357.1 | 7 | 2230 | |||
XM_017004869.1 | XP_016860358.1 | 7 | 2230 | |||
XM_017004870.1 | XP_016860359.1 | 7 | 2230 | |||
GenBank mRNA | AK303341.1 | - | 5 | 811 | ||
AK310628.1 | - | 5 | 815 | |||
AK310637.1 | - | 5 | 811 | |||
AL832287.1 | - | 1 | 13 | |||
BC039039.1 | - | 4 | 483 | 5168073 | ||
BC053878.1 | - | 5 | 805 | 5757161 | ||
L05148.1 | - | 5 | 831 |