Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034850.2 | NP_001030022.1 | 6 | 799 | ||
NM_019000.4 | NP_061873.2 | 4 | 675 | |||
XM_011514053.2 | XP_011512355.1 | 7 | 858 | |||
XM_011514054.2 | XP_011512356.1 | 5 | 809 | |||
XM_011514055.2 | XP_011512357.1 | 4 | 599 | |||
GenBank mRNA | AK024920.1 | - | 4 | 505 | ||
AK025808.1 | - | 6 | 726 | |||
AL832438.1 | - | 6 | 475 | |||
BC053326.1 | - | 6 | 723 | 5014868 | ||
BC073132.1 | - | 4 | 508 | 5760145 |