Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174936.3 | NP_777596.2 | 4 | 930 | ||
NR_110451.1 | 2 | 227 | ||||
GenBank mRNA | AK075365.1 | - | 4 | 859 | ||
AK122717.1 | - | 5 | 615 | |||
AK124635.1 | - | 1 | 1389 | |||
AK293870.1 | - | 5 | 910 | |||
AK297473.1 | - | 2 | 227 | |||
EF692496.1 | - | 4 | 568 |