Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 7 | 1248 | ||
NM_001321646.1 | NP_001308575.1 | 6 | 1221 | |||
NM_031466.7 | NP_113654.4 | 7 | 1423 | |||
XM_011517326.2 | XP_011515628.1 | 6 | 1396 | |||
XM_011517328.2 | XP_011515630.1 | 7 | 1423 | |||
XM_017013893.1 | XP_016869382.1 | 7 | 1423 | |||
GenBank mRNA | AB067469.1 | - | 7 | 1198 | ||
AK026394.1 | - | 7 | 1240 | |||
AK128755.1 | - | 7 | 1424 | |||
AL833973.1 | - | 5 | 524 | |||
AY190606.1 | - | 5 | 502 | |||
BC006206.2 | - | 5 | 644 | 3538806 |