Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015877.1 | NP_001015877.1 | 8 | 1023 | ||
NM_032335.3 | NP_115711.2 | 8 | 1026 | |||
NM_032458.2 | NP_115834.1 | 8 | 1023 | |||
GenBank mRNA | AB058726.1 | - | 8 | 909 | ||
AK290095.1 | - | 8 | 994 | |||
AK303369.1 | - | 7 | 886 | |||
AK303647.1 | - | 2 | 181 | |||
AY157622.1 | - | 8 | 1023 | |||
BC005994.1 | - | 8 | 965 | 4064169 | ||
CF242973.1 | - | 2 | 162 |