Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126132.2 | NP_001119604.1 | 12, 13 | 914 | ||
NM_001126133.2 | NP_001119605.1 | 11, 12 | 881 | |||
NM_001291774.1 | NP_001278703.1 | 12, 13 | 976 | |||
NM_003283.5 | NP_003274.3 | 12, 13 | 962 | |||
XM_011527246.2 | XP_011525548.1 | 11, 12 | 791 | |||
XM_017027186.1 | XP_016882675.1 | 13, 14 | 987 | |||
XM_017027187.1 | XP_016882676.1 | 12, 13 | 975 | |||
GenBank mRNA | AK291352.1 | - | 12, 13 | 766 | ||
AK308199.1 | - | 11, 12 | 733 | |||
AY762903.1 | - | 11, 12 | 709 | |||
AY762904.1 | - | 10, 11 | 676 | |||
BC010963.1 | - | 12, 13 | 767 | 4291828 | ||
BC022086.2 | - | 12, 13 | 815 | |||
BC034143.1 | - | 12, 13 | 766 | 4328089 | ||
BC107798.1 | - | 11, 12 | 736 | 4247379 | ||
BT019630.1 | - | 10, 11 | 676 | |||
F20363.1 | - | 1, 2 | 109 | |||
F34258.1 | - | 2, 3 | 148 | |||
M19308.1 | - | 11, 12 | 734 | |||
M19309.1 | - | 12, 13 | 814 | |||
S69208.1 | - | 12, 13 | 750 | |||
S69209.1 | - | 11, 12 | 717 |