Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018115.2 | NP_001018125.1 | 16 | 1488 | ||
NM_001319984.1 | NP_001306913.1 | 16 | 1560 | |||
NM_033084.4 | NP_149075.2 | 16 | 1488 | |||
GenBank mRNA | AF230336.1 | - | 16 | 1431 | ||
AF340183.1 | - | 16 | 1431 | |||
AK022613.1 | - | 9 | 890 | |||
AK307512.1 | - | 17 | 1461 |