Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014855.2 | NP_055670.1 | 17 | 2855 | ||
XM_017012864.1 | XP_016868353.1 | 16 | 2682 | |||
GenBank mRNA | AB007875.1 | - | 17 | 3079 | ||
AK092068.1 | - | 4 | 2318 | |||
BC008841.2 | - | 8 | 1523 | |||
BC014240.2 | - | 4 | 1046 | |||
BC037399.1 | - | 14 | 3057 | 5172434 |