Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 7 | 568 | ||
NM_152388.3 | NP_689601.2 | 7 | 733 | |||
GenBank mRNA | AB053301.1 | - | 7 | 735 | ||
AF212043.1 | - | 2 | 137 | |||
AK090601.1 | - | 7 | 426 | |||
AK303954.1 | - | 7 | 555 | |||
AK304395.1 | - | 5 | 257 | |||
BC029611.1 | - | 7 | 481 | 5304046 | ||
BQ219059.1 | - | 7 | 540 |