Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103511.1 | 5, 6 | 831 | |||
XM_006713783.3 | XP_006713846.1 | 4, 5 | 2235 | |||
XM_017007311.1 | XP_016862800.1 | 6, 7 | 2414 | |||
XM_017007312.1 | XP_016862801.1 | 5, 6 | 2469 | |||
XM_017007313.1 | XP_016862802.1 | 5, 6 | 2470 | |||
GenBank mRNA | AK292958.1 | - | 4, 5 | 495 | ||
AK310829.1 | - | 4, 5 | 486 | |||
AL136815.1 | - | 3, 4 | 262 | |||
BC024239.2 | - | 5, 6 | 601 | 4826873 | ||
BM705017.1 | - | 5, 6 | 452 |