Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 6 | 1781 | ||
NM_018344.5 | NP_060814.4 | 6 | 1775 | |||
NR_033413.1 | 5 | 1692 | ||||
NR_033414.1 | 4 | 1465 | ||||
XM_006717910.3 | XP_006717973.1 | 6 | 2167 | |||
XM_017016377.1 | XP_016871866.1 | 5 | 1989 | |||
XM_017016378.1 | XP_016871867.1 | 4 | 1765 | |||
GenBank mRNA | AK002022.1 | - | 5 | 1526 | ||
AX775915.1 | - | 6 | 1775 | |||
AX775917.1 | - | 6 | 1775 | |||
AX775919.1 | - | 6 | 1775 | |||
AX775921.1 | - | 6 | 1775 | |||
AY358686.1 | - | 6 | 1753 | |||
BC000223.1 | - | 3 | 1150 | |||
BC041575.1 | - | 5 | 1663 |