Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004429.4 | NP_004420.1 | 2 | 957 | ||
GenBank mRNA | BC016649.1 | - | 2 | 957 | 3867060 | |
BC052979.1 | - | 2 | 976 | 6164440 | ||
L37361.1 | - | 2 | 703 | |||
U09303.1 | - | 2 | 878 | |||
U09304.1 | - | 2 | 484 |