Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207346.2 | NP_997229.2 | 11 | 1836 | ||
XM_005257229.3 | XP_005257286.1 | 11 | 2023 | |||
GenBank mRNA | AK097583.1 | - | 9 | 1580 | ||
BC045551.1 | - | 12 | 1763 | |||
BC047793.1 | - | 11 | 1805 | 6500549 | ||
BC053643.1 | - | 11 | 1807 |