Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257229.3 | XP_005257286.1 | 8 | 1380 | ||
GenBank mRNA | AK094466.1 | - | 8 | 1293 |