Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317924.1 | NP_001304853.1 | 17 | 2160 | ||
NM_025132.3 | NP_079408.3 | 18 | 2276 | |||
XM_011513724.1 | XP_011512026.1 | 18 | 2186 | |||
XM_011513725.2 | XP_011512027.1 | 17 | 2141 | |||
XM_011513726.2 | XP_011512028.1 | 17 | 2049 | |||
XM_011513727.1 | XP_011512029.1 | 16 | 1969 | |||
XM_017008501.1 | XP_016863990.1 | 16 | 1969 | |||
GenBank mRNA | AB046858.1 | - | 8 | 1120 | ||
AK294730.1 | - | 17 | 2035 | |||
AK296112.1 | - | 2 | 250 | |||
AK304161.1 | - | 18 | 2149 | |||
AY029257.1 | - | 18 | 2276 |