Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533800
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
8, 9
1097
NM_001122764.1
NP_001116236.1
8, 9
1158
XM_005245291.3
XP_005245348.2
8, 9
1136
XM_005245295.3
XP_005245352.2
7, 8
984
XM_006711402.2
XP_006711465.2
8, 9
1103
XM_006711403.2
XP_006711466.2
8, 9
1092
XM_006711404.3
XP_006711467.2
8, 9
1040
XM_006711406.2
XP_006711469.2
8, 9
1013
XM_011509663.2
XP_011507965.1
8, 9
1223
XM_011509664.1
XP_011507966.1
8, 9
994
XM_011509665.2
XP_011507967.1
8, 9
1221
XM_011509666.2
XP_011507968.1
8, 9
1228
XM_011509667.2
XP_011507969.1
8, 9
1164
XM_011509668.2
XP_011507970.1
8, 9
964
XM_011509669.1
XP_011507971.1
8, 9
977
XM_011509670.2
XP_011507972.1
7, 8
1112
XM_011509671.1
XP_011507973.1
8, 9
994
XM_011509672.2
XP_011507974.1
7, 8
1103
XM_011509673.2
XP_011507975.1
6, 7
960
XM_011509675.2
XP_011507977.1
7, 8
1045
XM_011509676.1
XP_011507978.1
7, 8
1308
XM_011509677.1
XP_011507979.1
7, 8
1241
XM_011509678.1
XP_011507980.1
8, 9
1201
XM_011509679.1
XP_011507981.1
8, 9
1009
XM_011509681.1
XP_011507983.1
7, 8
926
XM_011509682.1
XP_011507984.1
7, 8
936
XM_017001559.1
XP_016857048.1
8, 9
1220
XM_017001560.1
XP_016857049.1
8, 9
1136
XM_017001561.1
XP_016857050.1
7, 8
1629
XM_017001562.1
XP_016857051.1
8, 9
1082
XM_017001563.1
XP_016857052.1
8, 9
1180
XM_017001564.1
XP_016857053.1
4, 5
698
XM_017001565.1
XP_016857054.1
8, 9
1082
XM_017001566.1
XP_016857055.1
8, 9
1191
XM_017001567.1
XP_016857056.1
7, 8
984
XM_017001568.1
XP_016857057.1
8, 9
1172
XM_017001569.1
XP_016857058.1
7, 8
926
XM_017001570.1
XP_016857059.1
5, 6
727
GenBank mRNA
AK094855.1
-
7, 8
930
AK302298.1
-
7, 8
968
BC002357.2
-
8, 9
1078
2821983
D38537.1
-
8, 9
1145
U26446.1
-
8, 9
1059
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline